1-(2,6-Difluoro-3-methylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183560-1G 183560-500MG 183560-5G 183560-25G
77466-244EA 415.09 USD
77466-244 77478-988 77489-222 77499-614
1-(2,6-Difluoro-3-methylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-(2,6-Difluoro-3-methylphenyl)-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₀H₅F₇O
MW: 274.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857747
CAS Number: 1939277-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR