1-(2-Ethoxy-5-methylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183534-1G 183534-500MG 183534-5G 183534-25G
77466-240EA 415.09 USD
77466-240 77478-984 77489-218 77499-610
1-(2-Ethoxy-5-methylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-(2-Ethoxy-5-methylphenyl)-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₁F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD34857730
CAS Number: 1988882-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR