2,2,3,3,3-Pentafluoro-1-(4-pentylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183526-1G 183526-500MG 183526-5G
77466-236EA 594.31 USD
77466-236 77478-980 77489-214
2,2,3,3,3-Pentafluoro-1-(4-pentylphenyl)-1-propanone ≥95%
2,2,3,3,3-Pentafluoro-1-(4-pentylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₄H₁₅F₅O
MW: 294.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28104076
CAS Number: 2173084-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR