1-[4-(Ethylthio)phenyl]-2,2,3,3,3-pentafluoro-1-propanone ≥97%

Supplier: AOBChem USA

Warning

183496-1G 183496-500MG 183496-5G 183496-25G
77466-228EA 422.3 USD
77466-228 77478-972 77489-206 77499-606
1-[4-(Ethylthio)phenyl]-2,2,3,3,3-pentafluoro-1-propanone ≥97%
1-[4-(Ethylthio)phenyl]-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₁H₉F₅OS
MW: 284.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15145656
CAS Number: 1443353-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR