2,2,3,3,3-Pentafluoro-1-(3-methoxyphenyl)-1-propanone ≥96%

Supplier: AOBChem USA

Warning

183472-1G 183472-500MG 183472-5G 183472-25G
77466-212EA 284.28 USD
77466-212 77478-956 77489-190 77499-604
2,2,3,3,3-Pentafluoro-1-(3-methoxyphenyl)-1-propanone ≥96%
2,2,3,3,3-Pentafluoro-1-(3-methoxyphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₀H₇F₅O₂
Storage Temperature: Ambient
MDL Number: MFCD15145648
CAS Number: 121592-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR