1-[1,1'-Biphenyl]-4-yl-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183469-1G 183469-500MG 183469-5G
77466-208EA 576.8 USD
77466-208 77478-952 77489-186
1-[1,1'-Biphenyl]-4-yl-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-[1,1'-Biphenyl]-4-yl-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₅H₉F₅O
MW: 300.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16615537
CAS Number: 1352226-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR