2-(3,5-Dibromo-4-methoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

18341-1G 18341-500MG 18341-5G
77466-200EA 471.74 USD
77466-200 77478-944 77489-178
2-(3,5-Dibromo-4-methoxyphenyl)-1,3-dioxolane ≥95%
2-(3,5-Dibromo-4-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀Br₂O₃
Storage Temperature: Ambient
MDL Number: MFCD31619397
CAS Number: 1629325-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR