3,5-Dimethyl-α,α-bis(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183236-1G 183236-500MG 183236-5G 183236-25G
77466-196EA 415.09 USD
77466-196 77478-940 77489-174 77499-594
3,5-Dimethyl-α,α-bis(1-methylethyl)benzenemethanol ≥95%
3,5-Dimethyl-α,α-bis(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O
Storage Temperature: Ambient
MDL Number: MFCD34857593
CAS Number: 1857579-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR