α,α-Bis(1-methylethyl)-4-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183202-1G 183202-500MG 183202-5G 183202-25G
77466-192EA 415.09 USD
77466-192 77478-936 77489-170 77499-590
α,α-Bis(1-methylethyl)-4-(methylthio)benzenemethanol ≥95%
α,α-Bis(1-methylethyl)-4-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂OS
MW: 238.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857560
CAS Number: 2141391-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR