2-Methyl-α,α-bis(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183194-1G 183194-500MG 183194-5G 183194-25G
77466-188EA 397.58 USD
77466-188 77478-932 77489-166 77499-586
2-Methyl-α,α-bis(1-methylethyl)benzenemethanol ≥95%
2-Methyl-α,α-bis(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD34857554
CAS Number: 898288-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR