α,3,5-Trimethyl-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183112-1G 183112-500MG 183112-5G
77466-184EA 648.9 USD
77466-184 77478-928 77489-162
α,3,5-Trimethyl-α-propylbenzenemethanol ≥95%
α,3,5-Trimethyl-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528595
CAS Number: 1379326-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR