α-Methyl-4-(2-methylpropyl)-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

183087-1G 183087-500MG 183087-5G 183087-25G
77466-180EA 352.26 USD
77466-180 77478-924 77489-158 77499-580
α-Methyl-4-(2-methylpropyl)-α-propylbenzenemethanol ≥95%
α-Methyl-4-(2-methylpropyl)-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O
Storage Temperature: Ambient
MDL Number: MFCD20526645
CAS Number: 1443327-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR