3-(1-Hydroxy-1-methylbutyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

183049-1G 183049-500MG 183049-5G
77466-176EA 447.02 USD
77466-176 77478-920 77489-154
3-(1-Hydroxy-1-methylbutyl)benzonitrile ≥97%
3-(1-Hydroxy-1-methylbutyl)benzonitrile

Precaución: For research use only

Formula: C₁₂H₁₅NO
MW: 189.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24217380
CAS Number: 1546518-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR