α,2,5-Trimethyl-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

182992-1G 182992-500MG 182992-5G
77466-172EA 668.47 USD
77466-172 77478-916 77489-150
α,2,5-Trimethyl-α-propylbenzenemethanol ≥95%
α,2,5-Trimethyl-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD16158864
CAS Number: 23124-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR