Tetrahydro-4-[3-(methylthio)phenyl]-2H-thiopyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

182913-1G 182913-500MG 182913-5G
77466-168EA 646.84 USD
77466-168 77478-912 77489-146
Tetrahydro-4-[3-(methylthio)phenyl]-2H-thiopyran-4-ol ≥95%
Tetrahydro-4-[3-(methylthio)phenyl]-2H-thiopyran-4-ol

Precaución: For research use only

Formula: C₁₂H₁₆OS₂
MW: 240.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31693978
CAS Number: 2150313-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR