1-Bromo-3,5-diisopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

18287-1G 18287-500MG 18287-5G 18287-25G
77466-164EA 117.42 USD
77466-164 77478-908 77489-142 77499-574
1-Bromo-3,5-diisopropylbenzene ≥95%
1-Bromo-3,5-diisopropylbenzene

Precaución: For research use only

Formula: C₁₂H₁₇Br
Storage Temperature: Ambient
MDL Number: MFCD12547791
CAS Number: 23058-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR