Benzenamine, 3-bromo-N-(diphenylmethylene)- ≥95%

Supplier: AOBChem USA

Warning

18280-1G 18280-500MG 18280-5G 18280-25G
77466-160EA 168.92 USD
77466-160 77478-904 77489-136 77499-568
Benzenamine, 3-bromo-N-(diphenylmethylene)- ≥95%
Benzenamine, 3-bromo-N-(diphenylmethylene)-

Precaución: For research use only

Formula: C₁₉H₁₄BrN
MW: 336.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28806013
CAS Number: 1020180-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR