5-Amino-2-bromo-4-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

182449-1G 182449-500MG 182449-5G 182449-25G
77466-156EA 427.45 USD
77466-156 77478-900 77489-132 77499-560
5-Amino-2-bromo-4-chlorobenzoic acid ≥97%
5-Amino-2-bromo-4-chlorobenzoic acid

Precaución: For research use only

Formula: C₇H₅BrClNO₂
MW: 250.4771 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547833
CAS Number: 1208077-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR