(3-Bromo-2,6-difluorophenyl)(cyclohexyl)methanol ≥97%

Supplier: AOBChem USA

Warning

182417-1G 182417-500MG 182417-5G 182417-25G
77466-152EA 415.09 USD
77466-152 77478-896 77489-128 77499-556
(3-Bromo-2,6-difluorophenyl)(cyclohexyl)methanol ≥97%
(3-Bromo-2,6-difluorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34857211
CAS Number: 1969534-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR