(2-Chloro-6-fluorophenyl)(cyclohexyl)methanol ≥96%

Supplier: AOBChem USA

Warning

182413-1G 182413-500MG 182413-5G 182413-25G
77466-148EA 339.9 USD
77466-148 77478-892 77489-124 77499-552
(2-Chloro-6-fluorophenyl)(cyclohexyl)methanol ≥96%
(2-Chloro-6-fluorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD16293497
CAS Number: 1226067-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR