(3-Bromophenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

182405-1G 182405-500MG 182405-5G
77466-144EA 449.08 USD
77466-144 77478-888 77489-120
(3-Bromophenyl)(cyclohexyl)methanol ≥95%
(3-Bromophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD12153603
CAS Number: 1183010-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR