(4-Chlorophenyl)(cyclohexyl)methanol ≥96%

Supplier: AOBChem USA

Warning

182399-1G 182399-500MG 182399-5G
77466-140EA 435.69 USD
77466-140 77478-884 77489-116
(4-Chlorophenyl)(cyclohexyl)methanol ≥96%
(4-Chlorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₇ClO
Storage Temperature: Ambient
MDL Number: MFCD01734779
CAS Number: 106165-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR