1-(5-Bromo-2-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥96%

Supplier: AOBChem USA

Warning

182393-1G 182393-500MG 182393-5G
77466-136EA 576.8 USD
77466-136 77478-880 77489-112
1-(5-Bromo-2-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥96%
1-(5-Bromo-2-fluorophenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₉H₃BrF₆O
Storage Temperature: Ambient
MDL Number: MFCD28107722
CAS Number: 1939277-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR