2,2,3,3,3-pentafluoro-1-(2-fluorophenyl)propan-1-one ≥97%

Supplier: AOBChem USA

Warning

182381-1G 182381-500MG 182381-5G
77466-128EA 594.31 USD
77466-128 77478-872 77489-104
2,2,3,3,3-pentafluoro-1-(2-fluorophenyl)propan-1-one ≥97%
2,2,3,3,3-pentafluoro-1-(2-fluorophenyl)propan-1-one

Precaución: For research use only

Formula: C₉H₄F₆O
MW: 242.1178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16615557
CAS Number: 1686161-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR