2-(5-Bromo-2-fluorophenyl)pentan-2-ol ≥97%

Supplier: AOBChem USA

Warning

182373-1G 182373-500MG 182373-5G
77466-124EA 447.02 USD
77466-124 77478-868 77489-098
2-(5-Bromo-2-fluorophenyl)pentan-2-ol ≥97%
2-(5-Bromo-2-fluorophenyl)pentan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₄BrFO
Storage Temperature: Ambient
MDL Number: MFCD23932765
CAS Number: 1525515-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR