2-(2-Chloro-6-fluorophenyl)pentan-2-ol ≥97%

Supplier: AOBChem USA

Warning

182369-1G 182369-500MG 182369-5G 182369-25G
77466-120EA 226.6 USD
77466-120 77478-864 77489-094 77499-542
2-(2-Chloro-6-fluorophenyl)pentan-2-ol ≥97%
2-(2-Chloro-6-fluorophenyl)pentan-2-ol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.6796 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857206
CAS Number: 1862852-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR