4-(2-hydroxypentan-2-yl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

182363-1G 182363-500MG 182363-5G
77466-116EA 447.02 USD
77466-116 77478-860 77489-090
4-(2-hydroxypentan-2-yl)benzonitrile ≥97%
4-(2-hydroxypentan-2-yl)benzonitrile

Precaución: For research use only

Formula: C₁₂H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD24217411
CAS Number: 1550630-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR