3-(5-Bromo-2-fluorophenyl)-2,4-dimethylpentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

182352-1G 182352-500MG 182352-5G 182352-25G
77466-112EA 415.09 USD
77466-112 77478-856 77489-084 77499-538
3-(5-Bromo-2-fluorophenyl)-2,4-dimethylpentan-3-ol ≥97%
3-(5-Bromo-2-fluorophenyl)-2,4-dimethylpentan-3-ol

Precaución: For research use only

Formula: C₁₃H₁₈BrFO
MW: 289.1838 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857205
CAS Number: 1870602-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR