3-(2-Chloro-6-fluorophenyl)-2,4-dimethylpentan-3-ol ≥95%

Supplier: AOBChem USA

Warning

182350-1G 182350-500MG 182350-5G 182350-25G
77466-108EA 415.09 USD
77466-108 77478-852 77489-080 77499-534
3-(2-Chloro-6-fluorophenyl)-2,4-dimethylpentan-3-ol ≥95%
3-(2-Chloro-6-fluorophenyl)-2,4-dimethylpentan-3-ol

Precaución: For research use only

Formula: C₁₃H₁₈ClFO
MW: 244.7328 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857203
CAS Number: 1877206-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR