3-(4-Fluoro-2-methylphenyl)-2,4-dimethylpentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

182347-1G 182347-500MG 182347-5G 182347-25G
77466-104EA 415.09 USD
77466-104 77478-848 77489-076 77499-530
3-(4-Fluoro-2-methylphenyl)-2,4-dimethylpentan-3-ol ≥97%
3-(4-Fluoro-2-methylphenyl)-2,4-dimethylpentan-3-ol

Precaución: For research use only

Formula: C₁₄H₂₁FO
MW: 224.3143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857200
CAS Number: 1878127-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR