3-(4-Chlorophenyl)-2,4-dimethylpentan-3-ol ≥95%

Supplier: AOBChem USA

Warning

182341-1G 182341-500MG 182341-5G 182341-25G
77466-100EA 226.6 USD
77466-100 77478-844 77489-072 77499-526
3-(4-Chlorophenyl)-2,4-dimethylpentan-3-ol ≥95%
3-(4-Chlorophenyl)-2,4-dimethylpentan-3-ol

Precaución: For research use only

Formula: C₁₃H₁₉ClO
MW: 226.7423 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857196
CAS Number: 104417-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR