4-Bromo-2-tert-butyl-aniline ≥95%

Supplier: AOBChem USA

Warning

18229-1G 18229-500MG 18229-5G 18229-25G
77466-092EA 90.64 USD
77466-092 77478-836 77489-062 77499-518
4-Bromo-2-tert-butyl-aniline ≥95%
4-Bromo-2-tert-butyl-aniline

Precaución: For research use only

Formula: C₁₀H₁₄BrN
Storage Temperature: Ambient
MDL Number: MFCD09965991
CAS Number: 850012-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR