5-Methyl-2-(methylthio)benzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

181660-1G 181660-500MG 181660-5G 181660-25G
77466-088EA 415.09 USD
77466-088 77478-832 77489-058 77499-512
5-Methyl-2-(methylthio)benzo[b]thiophene ≥95%
5-Methyl-2-(methylthio)benzo[b]thiophene

Precaución: For research use only

Formula: C₁₀H₁₀S₂
MW: 194.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28672425
CAS Number: 30489-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR