5-Methylbenzo[b]thiophene-2-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

181651-1G 181651-500MG 181651-5G 181651-25G
77466-084EA 162.74 USD
77466-084 77478-828 77489-054 77499-508
5-Methylbenzo[b]thiophene-2-carboxaldehyde ≥95%
5-Methylbenzo[b]thiophene-2-carboxaldehyde

Precaución: For research use only

Formula: C₁₀H₈OS
MW: 176.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865141
CAS Number: 27035-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR