2,2-Difluoro-α,α-Dimethyl-1,3-benzodioxole-4-methanol ≥95%

Supplier: AOBChem USA

Warning

181435-1G 181435-500MG 181435-5G
77466-076EA 591.22 USD
77466-076 77478-820 77489-044
2,2-Difluoro-α,α-Dimethyl-1,3-benzodioxole-4-methanol ≥95%
2,2-Difluoro-α,α-Dimethyl-1,3-benzodioxole-4-methanol

Precaución: For research use only

Formula: C₁₀H₁₀F₂O₃
MW: 216.18 g/mol
Storage Temperature: Ambient
CAS Number: 1891428-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR