1-(2,2-Difluoro-1,3-benzodioxol-4-yl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

181413-1G 181413-500MG 181413-5G
77466-072EA 597.4 USD
77466-072 77478-816 77489-040
1-(2,2-Difluoro-1,3-benzodioxol-4-yl)-1-propanone ≥95%
1-(2,2-Difluoro-1,3-benzodioxol-4-yl)-1-propanone

Precaución: For research use only

Formula: C₁₀H₈F₂O₃
MW: 214.17 g/mol
Storage Temperature: Ambient
CAS Number: 149740-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR