3-Ethoxy-α-propylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

181198-1G 181198-500MG 181198-5G 181198-25G
77466-068EA 341.96 USD
77466-068 77478-812 77489-036 77499-494
3-Ethoxy-α-propylbenzenemethanol ≥96%
3-Ethoxy-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521105
CAS Number: 1178920-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR