3-Fluoro-5-methyl-α-propylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

181149-1G 181149-500MG 181149-5G
77466-064EA 471.74 USD
77466-064 77478-808 77489-032
3-Fluoro-5-methyl-α-propylbenzenemethanol ≥96%
3-Fluoro-5-methyl-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD11521104
CAS Number: 1314924-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR