2-Fluoro-α-propyl-3-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

181119-1G 181119-500MG 181119-5G
77466-060EA 497.49 USD
77466-060 77478-804 77489-028
2-Fluoro-α-propyl-3-(trifluoromethyl)benzenemethanol ≥95%
2-Fluoro-α-propyl-3-(trifluoromethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₂F₄O
MW: 236.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23823176
CAS Number: 1508199-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR