4-Butyl-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

181048-1G 181048-500MG 181048-5G 181048-25G
77466-052EA 341.96 USD
77466-052 77478-796 77489-020 77499-486
4-Butyl-α-propylbenzenemethanol ≥95%
4-Butyl-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD11521076
CAS Number: 1226404-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR