3,5-Dimethyl-α-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

181003-1G 181003-500MG 181003-5G
77466-048EA 471.74 USD
77466-048 77478-792 77489-016
3,5-Dimethyl-α-propylbenzenemethanol ≥95%
3,5-Dimethyl-α-propylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD11521090
CAS Number: 691359-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR