α,α-Diethyl-3,5-Dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180951-1G 180951-500MG 180951-5G
77466-044EA 648.9 USD
77466-044 77478-788 77489-012
α,α-Diethyl-3,5-Dimethylbenzenemethanol ≥95%
α,α-Diethyl-3,5-Dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD12806372
CAS Number: 1178170-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR