α,α-Diethyl-3-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180918-1G 180918-500MG 180918-5G
77466-040EA 668.47 USD
77466-040 77478-784 77489-008
α,α-Diethyl-3-(methylthio)benzenemethanol ≥95%
α,α-Diethyl-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528657
CAS Number: 1379326-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR