α,α-Diethyl-4-(ethylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180909-1G 180909-500MG 180909-5G
77466-036EA 648.9 USD
77466-036 77478-780 77489-004
α,α-Diethyl-4-(ethylthio)benzenemethanol ≥95%
α,α-Diethyl-4-(ethylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528633
CAS Number: 1443305-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR