3-Ethoxy-α,α-diethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180903-1G 180903-500MG 180903-5G 180903-25G
77466-032EA 341.96 USD
77466-032 77478-776 77489-000 77499-480
3-Ethoxy-α,α-diethylbenzenemethanol ≥95%
3-Ethoxy-α,α-diethylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD20528658
CAS Number: 1443338-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR