α,α-Diethyl[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

180895-1G 180895-500MG 180895-5G 180895-25G
77466-028EA 415.09 USD
77466-028 77478-772 77488-996 77499-478
α,α-Diethyl[1,1'-biphenyl]-4-methanol ≥95%
α,α-Diethyl[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₇H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD21530088
CAS Number: 1484238-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR