α,α-Diethyl-4-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180860-1G 180860-500MG 180860-5G
77466-024EA 648.9 USD
77466-024 77478-768 77488-992
α,α-Diethyl-4-(methylthio)benzenemethanol ≥95%
α,α-Diethyl-4-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₈OS
MW: 210.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528634
CAS Number: 82955-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR