α,α-Diethyl-2-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

180836-1G 180836-500MG 180836-5G
77466-020EA 648.9 USD
77466-020 77478-764 77488-988
α,α-Diethyl-2-(1-methylethyl)benzenemethanol ≥95%
α,α-Diethyl-2-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
MW: 206.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526650
CAS Number: 858672-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR