(3-butoxy-2-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

180363-1G 180363-500MG 180363-5G 180363-25G
77466-016EA 415.09 USD
77466-016 77478-760 77488-982 77499-470
(3-butoxy-2-fluoro-4-methoxyphenyl)(methyl)sulfane ≥97%
(3-butoxy-2-fluoro-4-methoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₇FO₂S
MW: 244.3255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34856452
CAS Number: 2969427-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR