3-butoxy-2-fluoro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

180358-1G 180358-500MG 180358-5G 180358-25G
77466-012EA 415.09 USD
77466-012 77478-756 77488-978 77499-466
3-butoxy-2-fluoro-4-methoxybenzoic acid ≥97%
3-butoxy-2-fluoro-4-methoxybenzoic acid

Precaución: For research use only

Formula: C₁₂H₁₅FO₄
MW: 242.2435 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34856447
CAS Number: 2969418-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR